Learn More
Honeywell™ Turboshield Visor
Product Code. 15836665
Change view
Click to view available options
Lens Coating:
Anti-fog
Uncoated
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Coating unitSize
15836665 Uncoated Each
15846665 Anti-fog Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15836665 Supplier Honeywell™ Supplier No. 1031743
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
Toric-shaped polycarbonate visor for use with Honeywell™ Turboshield system.
  • Feel safer with 50% more chin coverage compared to traditional visors, without the need for bulky accessories
  • Visors can be easily removed with the new Turboshield Faceshield system using a simple push-button release.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE: 2, EC Certificate: C1519.2HON, EC Attestation: C1519.2HON, EN 166, EN 169, EN 170, EN 172
Lens Coating Uncoated
Description Dielectric
Lens Tint Clear
For Use With (Equipment) Turboshield headgear
For Use With (Application) Suitable face protection for most indoor work applications
Lens Material Polycarbonate