missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tween 20 blocking buffer, 1% in PBS (10X)

Product Code. 15464129
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
Unit Size:
1 L
2 L
500mL
Product Code. Quantity unitSize
15464129 1 L 1 L
15454129 500 mL 500mL
15474129 2 L 2 L
3 options
This item is not returnable. View return policy

Product Code. 15464129

Brand: Thermo Scientific Chemicals J61544.K2

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tween 20 blocking buffer solution is used as a washing agent in immunoassays, such as western blots and ELISAs. It helps to prevent non-specific antibody binding. The buffer is used for washes between each immunoreactions, to remove unbound immunological, and eventually for incubation solutions of immunoreagents to reduce unspecific background.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Blocking Buffer
Concentration 1% in PBS (10X)
Physical Form Viscous Liquid
Quantity 1 L

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.