missing translation for 'onlineSavingsMsg'
Learn More

Polysorbate 20, 100g, MP Biomedicals

Product Code. 11491622 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491622 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11491622 Supplier MP Biomedicals Supplier No. 04806576
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Polysorbate 20 is used as emulsifying agent for the preparation of stable oil-in-water emulsions in pharmaceuticals, cosmetics, cleaning compounds. Polysorbate 20 has been used in pre-extraction of membranes to remove peripheral proteins (used at 2% for extraction of membrane-bound proteins).

Polysorbate 20 has been used as a blocking agent for membrane based immunoassays at a typical concentration of 0.05%. It can be used for lysing mammalian cells at a concentration of 0.05 to 0.5%. It is a nonionic detergent widely used in biochemical applications.

Specifications

CAS 9005-64-5
Quantity 100 g
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
MDL Number MFCD00165986
Synonym Polyoxyethylenesorbitan monolaurate
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
Molecular Weight (g/mol) 522.68
PubChem CID 443314
Chemical Name or Material Polysorbate 20
Show More Show Less
compliance-icons
Product Identifier
  • POLYSORBATE 20
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Tween 20
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.