missing translation for 'onlineSavingsMsg'
Learn More

Tween 20™, Ultrapure, Thermo Scientific Chemicals

Product Code. 15825398 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 gal.
Packaging:
Plastic bottle
Unit Size:
4546mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15825398 500 mL Plastic bottle 500mL
15895178 1 gal. Plastic bottle 4546mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15825398 Supplier Thermo Scientific Supplier No. J20605.AP
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9005-64-5 | Molecular Formula: (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6 | Molecular Weight: 522.68 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyoxyethylene sorbitan monolaurate
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Physical Form Liquid
Boiling Point >100°C
Density 1.105
Quantity 500 mL
Flash Point >110°C
Packaging Plastic bottle
Specific Gravity 1.105
IUPAC Name 2-[2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy]ethyl dodecanoate
Grade Ultra Pure
Chemical Name or Material Tween 20™

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.