missing translation for 'onlineSavingsMsg'
Learn More

Tween 20™, Ultrapure, Thermo Scientific Chemicals

Product Code. 15895178 Shop All Thermo Scientific Products
Click to view available options
Quantity:
500 mL
1 gal.
Packaging:
Plastic bottle
Unit Size:
4546mL
500mL
This item is not returnable. View return policy

Product Code. 15895178

Brand: Thermo Scientific J20605.LO

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 9005-64-5
Molecular Formula (C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
Molecular Weight (g/mol) 522.68
MDL Number MFCD00165986
InChI Key HMFKFHLTUCJZJO-UHFFFAOYNA-N
Synonym Polyoxyethylene sorbitan monolaurate
PubChem CID 443314
SMILES CCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO

Specifications

Physical Form Liquid
Boiling Point >100°C
Quantity 1 gal.
Flash Point >110°C (230°F)
Packaging Plastic bottle
Specific Gravity 1.105
IUPAC Name 2-{2-[3,4-bis(2-hydroxyethoxy)oxolan-2-yl]-2-(2-hydroxyethoxy)ethoxy}ethyl dodecanoate
Grade Ultra Pure
Chemical Name or Material Tween 20™

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.