missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tween 20 washing buffer, 2% in PBS (20X)

Product Code. 15464029
Change view
Click to view available options
Quantity:
1 L
2 L
Unit Size:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15464029 1 L 1 L
15474029 2 L 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15464029 Supplier Thermo Scientific Chemicals Supplier No. J61419.K2
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PBS (Phosphate Buffered Saline) and Tween 20 are used as a wash buffer in immunolabeling techniques. Tween 20 is added to promote more effective washings, resulting in decreased non-specific background staining. Tween 20 is also known to promote reagent spreading in automated staining methodologies. 20X PBS Tween-20 is a space-saving stock solution that is ideal for preparing PBS-Tween (PBS-T) wash buffers for ELISA, Western and other immunoassays as well as a blocking buffer for plate-based assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Buffer
Concentration 2% in PBS (20X)
Physical Form Liquid
Quantity 1 L

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.