missing translation for 'onlineSavingsMsg'
Learn More

Tween™80, ∽75% oleic acid, MP Biomedicals™

Product Code. 11493601 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
0.5 L
1 L
4 L
Unit Size:
1 L
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11493601 1 L 1 L
11483601 0.5 L 500mL
11451981 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11493601 Supplier MP Biomedicals Supplier No. 0210317091
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 9005-70-3
Molecular Formula (C2H4O)x(C2H4O)z(C2H4O)y(C2H4O)wC42H76O7
Molecular Weight (g/mol) NaN
MDL Number MFCD01779641
InChI Key ZGEQUSWSYZZNAI-CLFAGFIQNA-N
Synonym Polysorbate 80, Polyoxyethylene sorbitan monooleate
SMILES CCCCCCCC\C=C/CCCCCCCC(=O)OCCOCC(OCCOC(=O)CCCCCCC\C=C/CCCCCCCC)C1OCC(OCCO)C1OCCO

Specifications

Color Yellow
Physical Form Liquid
Density 1.08g/mL
Quantity 1 L
Assay Percent Range ∽75%
Chemical Name or Material Tween 80
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.