missing translation for 'onlineSavingsMsg'
Learn More

Ohaus™ Two Block Analog Dry Block Heater HB2AL

Product Code. 15772699 Shop All Ohaus Products
Change view
Click to view available options
Controller Type:
Analog
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Controller Type Unit Size
15772699 Analog Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15772699 Supplier Ohaus™ Supplier No. 30392054
  • ONLINE EXCLUSIVE PRICE
    756.00 €
    710.01 € / Pack of 1
    Save 45.99 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

OHAUS™ Two Block Analog Dry Block Heater processes more samples while delivering precise and reliable results.

  • High wattage, constant-temperature Dry Block Heaters are economical, versatile and compact
  • Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures
  • Each of the five models accept separate interchangeable modular blocks (supplied separately) accommodating various sample enclosures such as microtubes, centrifuge tubes, vials, microplates, and PCR strips or tubes
  • Each block has a thermometer well for measuring block temperature
  • Anodised aluminum modular blocks provide superior temperature stability and heat transfer
  • Analogue controls
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance TUV
Controller Type Analog
Description Dry Block Heater, 2 Block, HB2AL, EU
Temperature Range (Metric) 5°C to 100°C (Low), 75°C to 150°C (High)
Type Dry Bath Block
Heat Up Time 50 min
Model HB2AL
No. of Blocks 2
Temperature Stability ±1.5°C
Temperature Uniformity ±0.1°C at 37°C
Uniformity ± 0.4°C at 37°C
Dimensions (L x W) 39.10 cm x 20.30 cm
Dimensions (L x W x H) 39.1 x 20.3 x 8.9 cm
Length (English) 15.40 in.
Length (Metric) 39.10 cm
Width (English) 8.00 in.
Width (Metric) 20.30 cm
Height (English) 3.50 in.
Height (Metric) 8.900 cm
Depth (English) 15.40 in.
Depth (Metric) 39.10 cm
Weight (Metric) 2.8 kg
Electrical Requirements 230V, 0.92A, 50/60Hz
Amperage 0.92A
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.