missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ Kel-F™ Fittings Assembled with PTFE FEP Tubing

Product Code. 11513194
Click to view available options
Thread Size:
1/4 in.-28
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11513194

Brand: Hamilton™ 88918

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Excellent for connecting syringes, equipment and other devices to Hamilton valves.

  • Manufactured from Kel-F™ PCTFE
  • Tubing gauge of 12
  • Tubing length of 304.8mm

Specifications

Type Cut End
Thread Size 1/4 in.-28
Description Gauge: 12
Overall Length (Metric) 304.8 mm
Material Kel-F
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.