missing translation for 'onlineSavingsMsg'
Learn More

Manutan Two tier commercial locker, blue/grey, 1800mm x 300mm x 500mm

Product Code. 12854649
Encompass_Preferred
This item is not returnable. View return policy
Product Code. 12854649 Supplier Manutan Supplier No. A330005
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for commercial, industrial, educational and leisure environments. 300mm deep lockers, fitted with hat shelf and two coat hooks. 500mm deep lockers, fitted with hat shelf, coat rail and single coat hook. Welded steel construction for added strength and rigidity. Supplied with cylinder lock and two keys per compartment. Pre-drilled for nesting side by side and/or back to back. Powder coated finish.

  • Blue door
  • Dimensions (H x W x D): 1800mm x 300mm x 500mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.