Learn More
Bel-Art™ SP Scienceware™ Two-Tier Polypropylene Test Tube Racks
Designed to withstand elevated water temperatures for incubation.
Brand: Bel-Art™ SP Scienceware™ F18513-2040
34.40 EUR valid until 2024-04-26
Use promo code "21699" to get your promotional price.
Additional Details : Weight : 0.50000kg
Description
Designed to withstand elevated water temperatures for incubation.
- Hold serum vials, RIA vials, T3 or T4 reaction vials, or any other vial or tube of same dimensions
- Won't rust or corrode
- Lightweight
- Autoclavable
- Polypropylene construction
- Includes endplate handles
Specifications
Tube Rack | |
4 x 10 | |
Tubes (16 to 20 mm) | |
40 | |
1 Each | |
254 mm | |
111 mm | |
89 mm | |
20 mm |
Integral endplate handles | |
Autoclavable | |
Polypropylene | |
Rectangle | |
254 x 111 x 89mm | |
10 in. | |
4.37 in. | |
3.5 in. |