missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ SP Scienceware™ Two-Tier Polypropylene Test Tube Racks
Designed to withstand elevated water temperatures for incubation.
40.70 €
Specifications
Includes | Integral endplate handles |
---|---|
Array | 4 x 10 |
Material | Polypropylene |
Positions | 40 |
Shape | Rectangle |
Product Code | Brand | Holds | Diameter (Metric) Opening | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Diameter (Metric) Opening | Price | Quantity & Availability | ||||
15500002
|
Bel-Art™ SP Scienceware™
F18513-1640 |
Tubes (13 to 16 mm) | 16 mm |
40.70 €
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
15375896
|
Bel-Art™ SP Scienceware™
F18513-2040 |
Tubes (16 to 20 mm) | 20 mm |
40.70 €
Each |
Estimated Shipment: 08-07-2024
Log in to see stock. |
||||
Description
Designed to withstand elevated water temperatures for incubation.
- Hold serum vials, RIA vials, T3 or T4 reaction vials, or any other vial or tube of same dimensions
- Won't rust or corrode
- Lightweight
- Autoclavable
- Polypropylene construction
- Includes endplate handles
Specifications
Integral endplate handles | |
Polypropylene | |
Rectangle | |
254 x 111 x 89mm | |
10 in. | |
4.37 in. | |
3.5 in. |
4 x 10 | |
40 | |
1 Each | |
254 mm | |
111 mm | |
89 mm |