missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies TWS119

Product Code. 17170232
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17170232 1 mg 1mg
17180232 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17170232 Supplier STEMCELL Technologies Supplier No. 73512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

WNT pathway activator; Inhibits GSK3β

  • TWS119 is a potent disubstituted pyrrolopyrimidine inhibitor of glycogen synthase kinase 3 beta (GSK3β) with an IC50 of 30 nM and Kd of 126 nM (Ding et al.) GSK3 is a serine/threonine kinase that is a key inhibitor of the WNT pathway; therefore TWS119 functions as a WNT pathway activator
  • Maintains the bi-potent, quiescent state in hepatic stellate cells of Wistar rats (Kordes et al.)
  • Induces differentiation of mouse embryonal carcinoma and embryonic stem cells to neurons (Ding et al.)
  • Induces production of T memory stem cell-like (T-SCM) cells from mouse or human CD8+ T cells with evidence of increased persistence, proliferation, and anti-tumor activity after adoptive transfer of mouse-derived T-SCM (Forget et al.; Gattinoni et al.)
  • Inhibits cell proliferation and induces apoptosis in human alveolar rhabdomyosarcoma cells (Zeng et al.)

Specifications

CAS 601514-19-6
Synonym GSK 3B Inhibitor XII
Quantity 1 mg
Molecular Formula C18H14N4O2
Purity ≥90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.