missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies TWS119
Description
- TWS119 is a potent disubstituted pyrrolopyrimidine inhibitor of glycogen synthase kinase 3 beta (GSK3β) with an IC50 of 30 nM and Kd of 126 nM (Ding et al.) GSK3 is a serine/threonine kinase that is a key inhibitor of the WNT pathway; therefore TWS119 functions as a WNT pathway activator
- Maintains the bi-potent, quiescent state in hepatic stellate cells of Wistar rats (Kordes et al.)
- Induces differentiation of mouse embryonal carcinoma and embryonic stem cells to neurons (Ding et al.)
- Induces production of T memory stem cell-like (T-SCM) cells from mouse or human CD8+ T cells with evidence of increased persistence, proliferation, and anti-tumor activity after adoptive transfer of mouse-derived T-SCM (Forget et al.; Gattinoni et al.)
- Inhibits cell proliferation and induces apoptosis in human alveolar rhabdomyosarcoma cells (Zeng et al.)
Specifications
Specifications
| CAS | 601514-19-6 |
| Synonym | GSK 3B Inhibitor XII |
| Quantity | 1 mg |
| Molecular Formula | C18H14N4O2 |
| Purity | ≥90% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?