Learn More
Thermo Scientific™ TX-100 Swinging Bucket Clinical Rotor
Efficiently process up to 16 x 5/7mL blood collection tubes per run.
Brand: Thermo Scientific™ 75005705
1620.49 EUR valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Code : NEW
Additional Details : Weight : 1.00000kg
Description
Efficiently process up to 16 x 5/7mL blood collection tubes per run with the Thermo Scientific™ TX-100 Swinging Bucket Clinical Rotor.
Ordering Information:
Includes unsealed carriers. Adapters are sold separately. Contact your sales representative for more information.
Specifications
TX-100 Swinging Bucket Clinical Rotor | |
16 x 15 mL | |
4,500 rpm | |
4258 | |
17 mm | |
90° |
Swinging Bucket Rotor | |
3,260 x g | |
144 mm | |
121 mm | |
Thermo Scientific Sorvall ST 8, Megafuge 8 and SL 8 Centrifuges |