Learn More
Thermo Scientific™ TX-750 Round Bucket Adapters
Add functionality to your rotor with round bucket adapters, which provide support for round-bottom and conical tubes and bottles, microtubes, and blood collection tubes.
Brand:  Thermo Scientific™ 75003719
Code : 51
Additional Details : Weight : 1.08000kg
 View more versions of this product
Product Code. 11874461
  • 790.00 € / Pack of 4
In Stock
to see stock.
Description
Description
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Specifications
Specifications
Round Bucket Adapter
84 x 15 mL
15mL Blood Collection Tube (17 x 125mm)
15 mL Blood Collection Tube
Round-Bottom and Conical Tube and Bottle, Microtube and Blood Collection Tube