missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 4000 S with Socks
GREENER_CHOICE

Product Code. 15919416 Shop All DuPont Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 15
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15919416 Small Pack of 15
15929416 Medium Pack of 15
15939416 Large Pack of 15
15949416 X-Large Pack of 15
15959416 2X-Large Pack of 15
15969416 3X-Large Pack of 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
MTO: Made to order terms & conditions apply. Working in Ex-Zones: Please take this into account for your risk-assessment that the attached socks may isolate the wearer. There is the possibility that the garment and wearer cannot by grounded via the shoes and other measures for grounding the garment and the wearer are required.The garment does not protect against ionizing radiation.This garment and/or fabric are not flame resistant and should not be used around heat, open flame, sparks or in potentially flammable environments.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15919416 Supplier DuPont™ Supplier No. D15193507
  • ONLINE EXCLUSIVE PRICE
    607.94 €
    572.63 € / Pack of 15
    Save 35.31 €
    5% Off
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
MTO: Made to order terms & conditions apply. Working in Ex-Zones: Please take this into account for your risk-assessment that the attached socks may isolate the wearer. There is the possibility that the garment and wearer cannot by grounded via the shoes and other measures for grounding the garment and the wearer are required.The garment does not protect against ionizing radiation.This garment and/or fabric are not flame resistant and should not be used around heat, open flame, sparks or in potentially flammable environments.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Tychem™ 4000 garments are made from a lightweight, supple and durable fabric. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.

  • DuPont™ Tychem™ 4000 S with socks. Hooded coverall with integrated socks and additional knee-length boot flaps. Available in white, in sizes SM to 3X. Elasticated face, wrists, waist and ankles for an optimal fit, thumb loops to prevent sleeves from riding up. Self-adhesive chin flap for tight seal of suit to face-mask.
    Tychem™ 4000 garments are made from a lightweight and durable fabric (<800g per garment) consisting of a chemical barrier film laminated to a heavy duty Tyvek™ substrate. They achieve a barrier to permeation by a broad range of inorganic and organic chemicals.
    Tychem™ 4000 S is ideal for chemical handling, environmental clean-up operations and emergency response. It is suitable for use in a variety of industries, including oil and gas, chemical engineering, and for use by hazardous material response teams and other emergency services.
  • For enhanced liquid protection, taping of outer cuff to glove is recommended. A double cuff is not a substitute for supplementary taping.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Tychem
Color White
Disposable Single-use
Ankle Style Elastic
Closure Type Double Zipper Front with Double Flaps
Seam Type Stitched, Taped
Sterility Non-sterile
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B
Resistance Chemicals, Liquid, Particle, Anti-static, Abrasion
Certifications/Compliance Certified according to Regulation (EU) 2016/425Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-BEN 14126 (barrier to infective agents), EN 1073-2 (protection againstradioactive contamination)Antistatic treatment (EN 1149-5) - on inside;
Description Tychem 4000 S with socks
No. of Pockets None
Size (English) Chest 33-36 in.
Size (Metric) Chest 84 to 92 cm
For Use With (Application) Chemical handling, environmental clean-up operations and emergency response.
Height (Metric) 162 to 170 cm
Weight (Metric) 124 g/m2
Quantity 15/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.