missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem™ 6000 AL TF640 00

Product Code. p-200008749
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 2
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 17906871

Brand: DuPont™ D15571100

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Tychem™ 6000 AL TF640 00 is a ventilated coverall with stitched and over-taped seams. It has a fully enclosed over hood with clear front and crown see through visor. Front closure with double flap hook and loop, single zipper closing to center, horizontal opening.

  • Ventilated coverall available in sizes small to 3X.
  • Cuff elastication and elastic thumb loop on inner sleeve opening.
  • Over boot assembly consisting of integrated dissipative inner boots attached to dissipative outerboots.
  • Gray.
    The protective system made of Tychem™ 6000 AL TF640 00 and the DuPont air-line belt, model 8000513914, helps protect the worker from contaminates.
  • The air distribution plenum made of Tychem™ 6000 fabric distributes cool and breathable air helping to reduce heat stress.
  • The simplified closure allows an easier donning procedure and can be taken off quickly thanks to an emergency egress strip. In addition to its top clear visor, this ventilated suit offers a non-PVC, highly chemical-resistant 180° panoramic view.
  • Tychem™ 6000 AL garments are made from a lightweight and durable fabric (<500g per garment) consisting of a proprietary barrier film laminated to Tyvek™ substrate.
  • They offer excellent protection against numerous hazardous industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
For Use With (Application) Protecting from Contaminents
Hood Type Fully Encapsulated
Material Tychem
Size Small
Closure Type Horizontal Zipper with Double Flap Hook and Loop
Disposable Single-use
Gender Unisex
Latex Free Yes
No. of Pockets None
Sterility Non-sterile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.