Learn More
DuPont™ Tychem™ 6000 F Plus with Dissipative Socks Model CHZ5
Product Code. 30305944
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 10
Product Code. 30305944
missing translation for 'mfr': DuPont™ D15546494
Hooded coverall with dissipative socks. Stitched and over-taped seams. Double cuffs. Knitted inner cuffs.
  • DuPont™ Tychem™ 6000 F Plus with dissipative socks, TFCHZ5TGY18.Hooded coverall. Stitched and over-taped seams. Double cuffs. Knitted inner cuffs. Thumb loops. Elastication at wrists, ankles, face, and waist. Double zippers and double flaps. Outer flap with adhesive tape. Integrated dissipative socks. Grey.
  • Tychem™ 6000 F garments are made from a lightweight and durable Tychem™ 6000 F fabric (<500g per garment) consisting of a proprietary barrier film laminated to a heavy duty Tyvek™ substrate. They offer excellent chemical permeation resistance to an extensive range of chemicals, helping to protect personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, biohazards and certain chemical warfare agents.
  • Tychem™ 6000 F garment are typically used for a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls with Socks
Size Small
Gender Unisex
Material HDPE
Color Gray
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Double
Neck Style Hooded
Hood Type Elastic
Closure Type Double Zipper Front with Double Flaps
Seam Type Stitched, Taped
Sterility Non-sterile
Resistance Liquid