missing translation for 'onlineSavingsMsg'
Learn More

Tyloxapol, MedChemExpress

Product Code. 16472309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472309 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16472309 Supplier Medchem Express Supplier No. HYB1068
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tyloxapol (Triton WR1339) is a nonionic liquid polymer of the alkyl aryl polyether alcohol type, used as a surface active stabilizer. Tyloxapol (Triton WR1339) is used to induce hyperlipidemia in animals.

Tyloxapol (Triton WR1339) is a nonionic liquid polymer of the alkyl aryl polyether alcohol type, used as a surface active stabilizer. Tyloxapol (Triton WR1339) is used to induce hyperlipidemia in animals.

Specifications

Chemical Name or Material Tyloxapol
Color Colorless
CAS 25301-02-4
Physical Form Viscous Liquid
Purity Grade Notes Research
Molecular Formula (C15H21O(C2H4O)m)n
Synonym Triton WR1339
Solubility Information H2O : 120 mg/mL (Need ultrasonic) ∣Ethanol : 100 mg/mL (Need ultrasonic) ∣DMSO : ≥ 38 mg/mL
Quantity 1 g
SMILES [H]OCCOC1=C(CC)C=C(C(CC(C)(C)C)(C)C)C=C1C.[6].[=].[<].[5].[8].[n].[m].[_].[=].[m].[n]
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.