missing translation for 'onlineSavingsMsg'
Learn More

Tyloxapol, Thermo Scientific Chemicals

Product Code. 10381023 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10381023 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10381023 Supplier Thermo Scientific Acros Supplier No. 422370050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25301-02-4

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25301-02-4
MDL Number MFCD00149002
Synonym Ethoxylated p-tert-octylphenol formaldehyde polymer

Specifications

Density 1.1000g/mL
pH 4.0 to 7.0 (5% aq. soln.)
Flash Point >113°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 10014
Specific Gravity 1.1
Solubility Information Solubility in water: soluble. Other solubilities: soluble in benzene, toluene, chloroform, carbon, tetrachloride, carbon disulfide, acetic acid
Physical Form Viscous Liquid
Chemical Name or Material Tyloxapol
Show More Show Less
compliance-icons
Product Identifier
  • Tyloxapol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Tyloxapol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.