missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 30 Combi Safety Storage Cabinets K-PHOENIX-30
Safe and approved storage of aggressive and flammable hazardous substances in working areas
Brand: asecos™ 3034200130345
Additional Details : Weight : 362.05920kg
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Two in one: 2 compartments (vertical seperated) allow the storage of flammable liquids and aggressive chemicals in one cabinet
- User-friendly and safe (folding door compartment): door open arrest system (standard), convenient and safe removal or storage of containers
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible) / cylinder lock
Specifications
1691 mm | |
Combi safety storage cabinets | |
532 mm | |
Powder-coated plain | |
1202 mm | |
598 x 1202 x 1968 mm | |
Left: 519 x 532 x 1691, mm, Right: 519 x 486 x 1691 mm | |
Tray shelf | |
1968 mm | |
K30.197.120.MV.FWAS | |
100 mm | |
Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated), 6 x tray shelf (panel melamine resin-coated/PP) | |
Safe and approved storage of aggressive and flammable hazardous substances in working areas | |
519 mm | |
Steel | |
Door open arrest system | |
380.50 kg | |
Light Gray | |
598 mm | |
Wing, Folding | |
9 | |
2 |