missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Robust construction and longevity: triple hinged door, cabinet body made of melamine resin and flame retardant special plates, safety elements assembled outside the storage compartment for increased protection against corrosion, doors with metal front as well as scratch-and impact-resistant surface, easy to clean
- All containers at a glance: door with large glass cutout allows a quick and easy overview of the stored containers – even with closed doors
- Easy handling with comfort: door stops in any position, self-closing in the event of fire
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy transport: integrated transport base with removable, optional base cover or adjusting aids with base cover
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Specifications
| Certifications/Compliance | Fire resistance 30 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
| Configuration | 6 Drawers |
| Depth (Metric) Exterior | 620 mm |
| Depth (Metric) Interior | 519 mm |
| Dimensions (D x W x H) Interior | 519 x 750 x 1626 mm |
| Dimensions (D x W x H) | 620 x 864 x 1947 mm |
| Door Style | Fixed Window, Left-hinged |
| For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
| Height (Metric) Exterior | 1947 mm |
| Height (Metric) Interior | 1626 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?