missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet Q-CLASSIC-90 Single Door

Safe and approved storage of flammable hazardous substances in working areas

Brand:  asecos™ 3007304134371

Additional Details : Weight : 287.80200kg

 View more versions of this product

Product Code. 15961636

  • 2620.85 € / Each
Estimated Shipment: 15-10-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Safe and approved storage of flammable hazardous substances in working areas
  • Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
  • Easy transport: integrated transport base with removable, optional base cover
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • EN 16121/16122 conformity
  • GS certified
  • CE compliant
Specifications

Specifications

1647 mm
3 x shelf, 1 x perforated insert, 1 x bottom collecting sump (sheet steel powder-coated)
Safe and approved storage of flammable hazardous substances in working areas
522 mm
Steel
615 x 599 x 1953 mm
522 x 450 x 1647 mm
Standard
1953 mm
Q90.195.060.R
90 mm
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Light gray doors
Safety storage cabinet
450 mm
Powder-coated textured
599 mm
314 kg
Anthracite Gray, Light Gray
615 mm
Swing, Right-hinged
3
1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers