missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Safe and approved storage of flammable hazardous substances in working areas
- Robust construction and longevity: triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch- and impact-resistant surface, easy to clean
- Effortless use: one-hand operation for double-door cabinets (AGT)
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: doors lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy transport: integrated transport base with removable, optional base cover
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system, spinning wheel in the exhaust grid as an indicator of sufficient technical ventilation
Specifications
Specifications
| Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
| Configuration | 6 x 5 L drawer |
| Depth (Metric) Exterior | 615 mm |
| Depth (Metric) Interior | 522 mm |
| Description | Warning Yellow doors |
| Dimensions (D x W x H) Interior | 522 x 450 x 1647 mm |
| Dimensions (D x W x H) | 615 x 599 x 1953 mm |
| Door Style | Swing, Right-hinged, Auto-close |
| For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
| Height (Metric) Exterior | 1953 mm |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?