missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet S-CLASSIC-90 with Door Open Arrest System (Yellow)

Product Code. 15984346 Shop All asecos Products
Change view
Click to view available options
Configuration:
2 Drawers
2 Shelves, 1 Perforated Insert (285 mm), 1 Bottom Collecting Sump
3 Drawers
3 Shelves, 1 Perforated Insert (185 mm), 1 Bottom Collecting Sump
3 Shelves, 1 Perforated Insert (370 mm), 1 Bottom Collecting Sump
3 Shelves, 1 Perforated Insert (370 mm), 1 Bottom Collecting Sump, 3 Tray Shelves, 1 Bottom Collecting Sump (PP), Vertical Dividing Wall
3 Shelves, 1 Perforated Insert (380 mm), 1 Bottom Collecting Sump
4 Drawers
6 Drawers
6 Drawers, 3 Tray Shelves, 1 Bottom Collecting Sump, Vertical Dividing Wall
Height (Metric) Exterior:
1298 mm
1968 mm
Width (Metric) Exterior:
896 mm
1196 mm
Unit Size:
Each
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Height (Metric) Exterior Width (Metric) Exterior unitSize
15984346 6 Drawers, 3 Tray Shelves, 1 Bottom Collecting Sump, Vertical Dividing Wall 1968 mm 1196 mm Each
15952876 2 Drawers 1298 mm 1196 mm Each
15962866 2 Shelves, 1 Perforated Insert (285 mm), 1 Bottom Collecting Sump 1298 mm 1196 mm Each
15962876 3 Drawers 1298 mm 1196 mm Each
15972866 3 Shelves, 1 Perforated Insert (185 mm), 1 Bottom Collecting Sump 1298 mm 1196 mm Each
15923036 3 Shelves, 1 Perforated Insert (370 mm), 1 Bottom Collecting Sump 1968 mm 896 mm Each
15902576 3 Shelves, 1 Perforated Insert (380 mm), 1 Bottom Collecting Sump 1968 mm 1196 mm Each
15974346 3 Shelves, 1 Perforated Insert (370 mm), 1 Bottom Collecting Sump, 3 Tray Shelves, 1 Bottom Collecting Sump (PP), Vertical Dividing Wall 1968 mm 1196 mm Each
15963036 4 Drawers 1968 mm 896 mm Each
15982576 6 Drawers 1968 mm 1196 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy

For indoor use only

Product Code. 15984346 Supplier asecos™ Supplier No. 3034800233849

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of flammable hazardous substances in working areas

  • Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
  • User-friendly and safe (model .WDAS): door open arrest system, convenient and safe removal or storage of containers
  • No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
  • Easy alignment: adjusting aids to compensate for uneven floor
  • Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration 6 Drawers, 3 Tray Shelves, 1 Bottom Collecting Sump, Vertical Dividing Wall
Depth (Metric) Exterior 616 mm
Depth (Metric) Interior 524 mm
Dimensions (D x W x H) Interior Left: 524 x 450 x 1742 mm, Right: 524 mmx 450 x 1742 mm
Dimensions (D x W x H) 616 x 1196 x 1968 mm
Door Style Wing
For Use With (Application) Safe and approved storage of flammable hazardous substances in working areas
Height (Metric) Exterior 1968 mm
Height (Metric) Interior 1742 mm
Includes Door open arrest system
Model S90.196.120.MV.WDAS
No. of Doors 2
No. of Shelves 3
Shelf Type Tray shelf
Surface Coating Powder-coated plain
Weight (Metric) 514.50 kg
Width (Metric) Exterior 1196 mm
Width (Metric) Interior 450 mm
Color Warning Yellow
Material Steel
Product Type Flammables Storage Cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.