missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX Vol.2-90 with Integrated AGT
Safe and approved storage of flammable hazardous substances in working areas
3414.26 € - 4499.17 €
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Depth (Metric) Exterior | 616 mm |
Depth (Metric) Interior | 524 mm |
Description | One-hand operation for double-door cabinets (AGT) |
Dimensions (D x W x H) | 616 x 1196 x 1968 mm |
Alert:
For indoor use only
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Configuration | Color | Price | Quantity & Availability | ||||
15556855
|
asecos™
3011900133715 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
3414.26 €
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15982686
|
asecos™
3011900233715 |
3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow |
3414.26 €
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15942666
|
asecos™
3011900133719 |
3 x tray shelf, 1 x perforated insert (365 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
3616.72 €
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15922696
|
asecos™
3011900233719 |
3 x tray shelf, 1 x perforated insert (365 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
3616.72 €
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15992666
|
asecos™
3011900133724 |
4 x drawer (sheet steel powder-coated) | Light Gray |
4056.09 €
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15972696
|
asecos™
3011900233724 |
4 x drawer (sheet steel powder-coated) | Warning Yellow |
4056.09 €
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15992696
|
asecos™
3011900233726 |
6 x drawer (sheet steel powder-coated) | Warning Yellow |
4499.17 €
Each |
Estimated Shipment: 15-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding doors: less required space in front of the cabinet while doors are open
- Effortless use: one-hand operation for double-door cabinets (AGT)
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested doors after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: doors lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
524 mm | |
616 x 1196 x 1968 mm | |
Steel | |
Folding, Auto-close | |
1968 mm | |
S90.196.120.FDAC | |
Powder-coated plain | |
1050 mm |
616 mm | |
One-hand operation for double-door cabinets (AGT) | |
524 x 1050 x 1742 mm | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm | |
2 | |
1196 mm |