missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos™ Type 90 Safety Storage Cabinet S-PHOENIX Vol.2-90 Left-Hinged
Safe and approved storage of flammable hazardous substances in working areas
2595.94 € - 3403.66 €
Specifications
Certifications/Compliance | Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |
---|---|
Material | Steel |
Product Type | Safety storage cabinet |
Door Style | Folding, Left-hinged, Auto-close |
For Use With (Application) | Safe and approved storage of flammable hazardous substances in working areas |
Alert:
For indoor use only
Product Code | Brand | Depth (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (Metric) Exterior | Configuration | Color | Price | Quantity & Availability | |||
15993576
|
asecos™
3023500133659 |
616 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
2595.94 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15903616
|
asecos™
3023500233659 |
616 mm | 3 x shelf, 1 x perforated insert (380 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Yellow |
2595.94 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15933586
|
asecos™
3023500133663 |
616 mm | 3 x tray shelf, 1 x perforated insert (320 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Light Gray |
2797.34 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15943616
|
asecos™
3023500233663 |
616 mm | 3 x tray shelf, 1 x perforated insert (320 mm), 1 x bottom collecting sump (sheet steel powder-coated) | Warning Yellow |
2797.34 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15943586
|
asecos™
3023500133681 |
616 mm | 4 x drawer (sheet steel powder-coated) | Light Gray |
2946.80 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15953616
|
asecos™
3023500233681 |
616 mm | 4 x drawer (sheet steel powder-coated) | Warning Yellow |
2946.80 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15953586
|
asecos™
3023500133683 |
616 mm | 6 x drawer (sheet steel powder-coated) | Light Gray |
3226.64 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15973616
|
asecos™
3023500233683 |
616 mm | 6 x drawer (sheet steel powder-coated) | Warning Yellow |
3226.64 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15934216
|
asecos™
3031500133695 |
749 mm | 4 x drawer (sheet steel powder-coated) | Light Gray |
3083.54 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15954226
|
asecos™
3031500233695 |
749 mm | 4 x drawer (sheet steel powder-coated) | Warning Yellow |
3083.54 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15954216
|
asecos™
3031500133697 |
749 mm | 6 x drawer (sheet steel powder-coated) | Light Gray |
3403.66 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15974226
|
asecos™
3031500233697 |
749 mm | 6 x drawer (sheet steel powder-coated) | Warning Yellow |
3403.66 €
Each |
Estimated Shipment: 14-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
- Robust construction and longevity: triple hinged door, body made of powder-coated sheet steel, safety elements assembled outside the cabinet for increased protection against corrosion
- Space saving folding door: less required space in front of the cabinet while door is open
- User-friendly and safe: door open arrest system (standard), convenient and safe removal or storage of containers
- Always closed: serial automatic door closing (TSA) releases arrested door after approx. 60 seconds; acoustic and visual signal before closing
- No unauthorised use: door lockable with profile cylinder (integration in an existing locking system possible)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Specifications
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant | |
Safety storage cabinet | |
Safe and approved storage of flammable hazardous substances in working areas | |
1742 mm | |
Powder-coated plain | |
450 mm |
Steel | |
Folding, Left-hinged, Auto-close | |
1968 mm | |
1 | |
596 mm |