missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Cabinet V-CLASSIC-90
BIOPHARMAGREENER_CHOICE

Product Code. p-7250628 Shop All asecos Products
Change view
Click to view available options
Includes:
Tray Shelf
Tray Shelf w/ Storage Box
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
17805814 Tray Shelf Each
17815814 Tray Shelf w/ Storage Box Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy

For indoor use only

Product Code. 17805814 Supplier asecos™ Supplier No. 4028500540627

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For indoor use only

The MULTIRISK cabinet from asecos™ for safe, flexible, and energetically sustainable storage of various hazardous materials in just one cabinet.

Function / construction

  • Special fire resistant construction: The special, completely fire-resistant construction of the Type 90 MULTIRISK cabinet offers, for the first time, the possibility of fire-protected storage of corrosive and toxic hazardous substances
  • The cabinet thus not only ensures the safe storage of different hazardous materials in daily operations, but also provides maximum protection for users and rescue forces in case of a fire
  • Locking: To protect against unauthorised use, the cabinet can be securely locked
  • The integrated red/green indicator quickly shows the locking status
  • The cabinet can be integrated into existing locking systems by replacing the profile cylinder on site
  • Comfortable overview and access: The cabinet user is entirely unrestricted in deciding which types of hazardous materials are to be stored in the two storage compartments
  • Both vertical drawers can be used equally thanks to the ventilation concept and uniform interior equipment
  • Metal-free storage levels: Each storage level is equipped with an tray shelf made of chemical-resistant PE and can be adjusted tool-free in height at any time by the customer
  • The polyethylene trays are easy to remove, e.g. for cleaning or in case of leakage
  • Optimal integration and use of space: The space-saving storage solution with high storage volume is optimally suited for installation next to fume cupboards with a width of only 60 cm and a depth of 86 cm deep
  • Technical ventilation: Hazardous material vapours are captured at each storage level within the cabinet and can be extracted directly via the integrated air ducts ready for conneciton (NW 75) to a technical exhaust system
  • The possible formation of explosive atmospheres or harmful vapours is safely prevented

Certificates

  • Fire resistance 90 minutes
  • Type tested in accordance with EN 14470-1
  • DIN EN 16121/16122 conformity
  • GS certified
  • CE compliant

Specifications

Certifications/Compliance Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant
Configuration Per Door: 4 x tray shelf, 4 x perforated insert (PP), 1 x bottom collecting sump (polyethylene)
Depth (English) Interior 306.69 in.
Dimensions (D x W x H) Interior 779 x 506 x 1750 mm
Dimensions (D x W x H) 860 x 600 x 1966 mm
Door Style Vertical Drawer
For Use With (Application) For the unrestricted storage of a variety of hazardous materials in work areas
Height (English) Interior 688.98 in.
Includes Tray Shelf
Load Bearing Capacity 30 kg
Model V90.196.060.MV.2VD:0012
No. of Doors 2
No. of Shelves 8
Shelf Type Tray Shelf
Weight (English) 1005.31 lb.
Weight (Metric) 456 kg
Width (English) Interior 199.21 in.
Color White
Depth (English) Exterior 338.58 in.
Height (English) Exterior 774.02 in.
Height (Metric) Opening 1750 mm
Material Gypsum
Product Type Flammables Storage Cabinet
Width (English) Exterior 236.22 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.