missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos Type 90 Safety Storage Cabinet V-MOVE-90
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos 3508604635103
Alert:
For indoor use only
Description
- Robust construction and longevity: robust, high-quality, scratch-resistant outer body with powder-coated surface
- Electronic opening and closing: the vertical drawers open and close at the push of a button
- Perfect access: the vertical drawers provide an overview and simple access to the stored containers
- Always closed: serial automatic door closing (TSA) after approx. 60 seconds with acoustic and visual signal before drawer closing
- No unauthorised use: vertical drawers lockable with cylinder locking (integration in an existing locking system possible) and locking state indicator (red/green)
- Easy alignment: adjusting aids to compensate for uneven floor
- Ventilation: integrated air ducts ready for connection (DN 75) to a technical exhaust system
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
701 mm | |
77.40 in. | |
Gypsum | |
860 x 819 x 1966 mm | |
33.86 in. | |
701 x 775 x 1750 mm | |
1226 lb. | |
10 | |
68.90 in. | |
Safety Storage Cabinet | |
32.24 in. | |
Red, Anthracite Gray | |
1966 mm | |
V90.196.081.VDAC:0013 | |
2 |
775 mm | |
Textured | |
Right: (5) shelves, (1) perforated insert, (1) bottom collecting sump; Left: (5) shelves, (1) perforated insert, (1) bottom collecting sump (sheet steel powder-coated) | |
556 kg | |
30.51 in. | |
860 mm | |
50 kg | |
27.60 in. | |
1750 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
819 mm | |
Standard | |
Auto-close | |
235/250/250/250/285/260 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, GS certified, CE compliant |