missing translation for 'onlineSavingsMsg'
Learn More
Learn More
asecos Type 90 Safety Storage Under Bench Cabinet UB-T-90
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos 3591500130488
Additional Details : Weight : 8.00000kg
Alert:
For indoor use only
Description
- Robust construction and longevity: body made of powder-coated sheet steel
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Certificates
- Fire resistance: 90 minutes
- Type tested in accordance with EN 14470-1
- DIN EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
470 mm | |
31.50 in. | |
Gypsum | |
600 x 593 x 800 mm | |
23.62 in. | |
470 x 482 x 702 mm | |
305 lb. | |
1 | |
27.64 in. | |
Under Bench Safety Storage Cabinet | |
23.35 in. | |
Light Gray | |
800 mm | |
UB90.080.059.060.TR | |
1 |
482 mm | |
Plain | |
(1) perforated insert, (1) bottom collecting sump (stainless steel) | |
139 kg | |
18.98 in. | |
600 mm | |
75 kg | |
18.50 in. | |
702 mm | |
Safe and approved storage of flammable hazardous substances in working areas | |
593 mm | |
Perforated insert | |
Right-hinged, Wing | |
600 mm | |
Fire resistance 90 minutes, type tested in accordance with EN 14470-1, DIN EN 16121/16122 conformity, CE compliant |