Learn More
asecos™ Type 90 Safety Storage Under Bench Cabinet UB-T-90
Safe and approved storage of flammable hazardous substances in working areas
Brand: asecos™ 3265800135378
Code : NEW
Additional Details : Weight : 157.50000kg
Alert:
For indoor use only
Description
Safe and approved storage of flammable hazardous substances in working areas
Function / construction
- Robust construction and longevity: body made of powder-coated sheet steel
- Easy handling with comfort: doors stop in any position, self-closing in the event of fire
- User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
- No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
- Mobile: castors with plinth (optionally available), easy moving of the cabinet
- Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system
Certificates
- Fire resistance 90 minutes
- type tested in accordance with EN 14470-1
- EN 16121/16122 conformity
- GS certified
- CE compliant
Specifications
702 mm | |
Storage of flammable liquids | |
482 mm | |
Steel | |
143 kg | |
Light Gray | |
EN 14470-1 with a fire resistance of 90 minutes |
One pullout drawer, perforated metal plate insert and one second level drawer | |
470 mm | |
Powder-coated | |
593 mm | |
482 x 470 x 702 mm | |
800 mm |