missing translation for 'onlineSavingsMsg'
Learn More

asecos™ Type 90 Safety Storage Under Bench Cabinet UB-T-90

Product Code. 15843447 Shop All asecos Products
Change view
Click to view available options
Configuration:
1 Pull-out Drawer, 1 Perforated Insert, 1 Second Level Drawer
2 Drawers, 2 Perforated Inserts
Dimensions (D x W x H) Interior:
482 x 470 x 702 mm
482 x 979 x 702 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Configuration Dimensions (D x W x H) Interior unitSize
15843447 1 Pull-out Drawer, 1 Perforated Insert, 1 Second Level Drawer 482 x 470 x 702 mm Each
15365569 2 Drawers, 2 Perforated Inserts 482 x 979 x 702 mm Each
15365579 1 Pull-out Drawer, 1 Perforated Insert, 1 Second Level Drawer 482 x 470 x 702 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy

For indoor use only

Product Code. 15843447 Supplier asecos™ Supplier No. 3265800135378

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For indoor use only

Safe and approved storage of flammable hazardous substances in working areas

Safe and approved storage of flammable hazardous substances in working areas

Function / construction

  • Robust construction and longevity: body made of powder-coated sheet steel
  • Easy handling with comfort: doors stop in any position, self-closing in the event of fire
  • User-friendly: access to all containers, 93° opening of the wing door, interior of the cabinet completely visible
  • No unauthorised use: doors lockable with cylinder lock and locking state indicator (red/green)
  • Mobile: castors with plinth (optionally available), easy moving of the cabinet
  • Ventilation: integrated air ducts ready for connection (DN 50) to a technical exhaust system

Certificates

  • Fire resistance 90 minutes
  • type tested in accordance with EN 14470-1
  • EN 16121/16122 conformity
  • GS certified
  • CE compliant

Compliance

DIN EN 14470-1 - Fire resistance of 90 minutes

Specifications

Certifications/Compliance EN 14470-1 with a fire resistance of 90 minutes
Configuration 1 Pull-out Drawer, 1 Perforated Insert, 1 Second Level Drawer
Depth (Metric) Interior 482 mm
Description Drawer: 50 kg/25 L, Insert: 75 kg, Second level drawer: 25 kg/3 L, Maximum load: 300 kg
Dimensions (D x W x H) Interior 482 x 470 x 702 mm
Dimensions (D x W x H) 600 x 593 x 800 mm
Door Style Locking
For Use With (Application) Storage of flammable liquids
Height (Metric) Exterior 800 mm
Height (Metric) Interior 702 mm
Load Bearing Capacity 300 kg
No. of Doors 1
Surface Coating Powder-coated
Weight (Metric) 143 kg
Width (Metric) Exterior 593 mm
Width (Metric) Interior 470 mm
Color Light Gray
Material Steel
Product Type Safety storage cabinet
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.