missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Tyr-Gly-Gly-Phe-Met-OH

Product Code. 16469196
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16469196 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16469196 Supplier MedChemExpress Supplier No. HYP0073
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tyr-Gly-Gly-Phe-Met-OH regulates human immune function and inhibits tumor growth via binding to the opioid receptor.

Specifications

Purity 98.07%
CAS 58569-55-4
Content And Storage Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month
For Use With (Application) Cancer-programmed cell death
Molecular Weight (g/mol) 573.66 g/mol
Product Type Peptides
Quantity 100 mg
Shipping Condition Room temperature
Solubility DMSO : ≥ 40 mg/mL (69.73 mM) ∣H2O : 6.67 mg/mL (11.63 mM∣Need ultrasonic)
Synonym Met-Enkephalin∣ Methionine enkephalin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.