missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Tyramide Conjugates

Product Code. 15595055
Change view
Click to view available options
Conjugate:
Alexa Fluor™ 350
Alexa Fluor™ 488
Alexa Fluor™ 546
Alexa Fluor™ 555
Alexa Fluor™ 568
Alexa Fluor™ 594
Alexa Fluor™ 647
Alexa Fluor™ Plus 750
Biotin-XX
Unit Size:
0.5mg
Each
Pack of 150
Excitation/Emission:
347/442 nm
495/519 nm
555/565 nm
556/573 nm
579/604 nm
591/617 nm
650/668 nm
750/790 nm
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Conjugate Unit Size Excitation/Emission
15595055 Biotin-XX Each -
15651902 Alexa Fluor™ 350 Pack of 150 347/442 nm
15542414 Alexa Fluor™ 488 Each 495/519 nm
15661902 Alexa Fluor™ 546 Pack of 150 556/573 nm
15671902 Alexa Fluor™ 555 Pack of 150 555/565 nm
15681902 Alexa Fluor™ 568 Pack of 150 579/604 nm
15539135 Alexa Fluor™ 594 Each 591/617 nm
15691902 Alexa Fluor™ 647 Pack of 150 650/668 nm
17836578 Alexa Fluor™ Plus 750 0.5mg 750/790 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 15595055 Supplier Invitrogen™ Supplier No. B40951
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Fisher Scientific offers a wide variety of bright Alexa Fluor™, Alexa Fluor™ Plus or Biotin XX tyramide conjugates. These tyramide conjugates are used in tyramide signal amplification (TSA), also called catalyzed reporter deposition (CARD).

Thermo Fisher Scientific offers a wide variety of bright Alexa Fluor™, Alexa Fluor™ Plus or Biotin XX tyramide conjugates. These tyramide conjugates are used in tyramide signal amplification (TSA), also called catalyzed reporter deposition (CARD). TSA is used to amplify signals up to 200 fold versus standard imaging methods. This reagent can be used with any antibody conjugated to HRP for tyramide signal amplification to detect low abundance targets. Up to seven-plex is possible in multiplexable fluorescent imaging.

These tyramide reagents are used in tyramide signal amplification (TSA), also called catalyzed reporter deposition (CARD). TSA is used to amplify signal up to 200-fold versus standard imaging methods. These reagents are provided stand-alone, but are also available as part of our SuperBoost™ tyramide signal amplification kits (available separately). They can be used with any antibody conjugated to HRP for tyramide signal amplification for detection of low abundance targets. Up to seven-plex is possible in multiplex fluorescent immunocytochemistry (ICC), immunohistochemistry (IHC), and in situ hybridization (ISH).

Features include:
• Superior sensitivity for detection of low-level or hard-to-detect targets by fluorescent imaging
• Simple protocol and detection using standard filters
• Suitable for high-resolution multiplex images—co-label with TO-PRO-3, secondary antibodies, and other SuperBoost kits
• Requires 10–100 times less primary antibody then standard ICC/IHC/ISH experiments

Tyramide reagents, when used as directed, can increase sensitivity up to 200 times over standard imaging methods. These reagents help sharpen your results for clear visibility into critical areas that standard imaging methods fail to reveal. They are simple to use and easily adapted to standard ICC, IHC, or FISH experimental protocols, using any cell or tissue type. Cells labeled using a tyramide reagent can be imaged using any type of microscope, producing high-resolution multiplex images.

Dissolve the Alexa Fluor™ or Biotin-XX tyramide reagent in 150 μL of DMSO to prepare 100X stock. Tyramide may coat the sides of the vial so invert the vial several times to ensure all reagent is dissolved in solution.

TRUSTED_SUSTAINABILITY

Specifications

Clonality None
Quantity 1 vial, 150 slides
For Use With (Application) Immunohistochemistry (IHC), Immunofluorescence (IF), Immunocytochemistry (ICC), ISH
Conjugate Biotin-XX
Content And Storage • 1 vial Biotin-XX Tyramide Reagent sufficient for 150 slides

Store at 2°C to 8°C. When stored as directed, the product is stable for 6 months after receiving.
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.