missing translation for 'onlineSavingsMsg'
Learn More

Tyramine, 97%

Product Code. 10666532 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10666532 25 g Glass bottle 25g
10581191 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10666532 Supplier Thermo Scientific Acros Supplier No. 140610250
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 51-67-2 | C8H11NO | 137.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-67-2
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00008193
InChI Key DZGWFCGJZKJUFP-UHFFFAOYSA-N
Synonym tyramine, 4-2-aminoethyl phenol, 4-hydroxyphenethylamine, p-tyramine, 2-4-hydroxyphenyl ethylamine, uteramine, tyramin, tyrosamine, tocosine, 4-hydroxyphenylethylamine
PubChem CID 5610
ChEBI CHEBI:15760
IUPAC Name 4-(2-aminoethyl)phenol
SMILES C1=CC(=CC=C1CCN)O

Specifications

Melting Point 158.0°C to 163.0°C
Color Brown to Yellow
Boiling Point 175.0°C to 181.0°C (8.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula HOC6H4CH2CH2NH2
Quantity 25 g
Beilstein 13, 625
Merck Index 15, 10016
Solubility Information Solubility in water: 1g/95mL (15°C). Other solubilities: 1g/10mL boiling alcohol, sparingly soluble in benzene and xylene, soluble in ethanol
Formula Weight 137.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Tyramine, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Tyramine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.