missing translation for 'onlineSavingsMsg'
Learn More

Tyramine hydrochloride, 99%

Product Code. 10036000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10036000 5 g Glass bottle 5g
10587942 25 g Glass bottle 25g
10423575 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10036000 Supplier Thermo Scientific Acros Supplier No. 422380050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tyramine hydrochloride, CAS # 60-19-5, is the hydrochloride salt of tyramine, a biogenic amine derived from the amino acid tyrosine. | CAS: 60-19-5 | C8H12ClNO | 173.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Tyramine is a naturally occurring monoamine that is derived from the decarboxylation of the amino acid tyrosine utilized in human and invertebrate nervous systems
  • Tyramine is synthesized and released from neurons and acts as a neuromodulator upon binding to tyramine receptors
  • Tyramine has been found to impact the immune system and cardiovascular system in humans

Applications

  • Tyramine may be used to form tyramide conjugate, which can be used as a signal amplification reagent in immunofluorescence assays
  • Tyramine-hyaluronic acid conjugates can be used to form biodegradable hydrogels, with utility in drug delivery and tissue engineering
  • Tyramine is useful for laboratory studies of neuronal stimulation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-19-5
Molecular Formula C8H12ClNO
Molecular Weight (g/mol) 173.64
MDL Number MFCD00012901
InChI Key RNISDHSYKZAWOK-UHFFFAOYSA-N
Synonym tyramine hydrochloride, 4-2-aminoethyl phenol hydrochloride, 4-hydroxyphenethylamine hydrochloride, tyrosamine hydrochloride, tyramine monochloride, p-tyramine hydrochloride, phenol, 4-2-aminoethyl-, hydrochloride, p-tryamine hydrochloride, tyramine hcl, unii-z5kdh3h147
PubChem CID 66449
SMILES [H+].[Cl-].NCCC1=CC=C(O)C=C1

Specifications

Melting Point 271°C to 274°C
Color White
Boiling Point 269°C
Loss on Drying 0.5% max. (105°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98.5 to 101.5% (on dry substance) (Argentometry)
Packaging Glass bottle
Linear Formula HOC6H4CH2CH2NH2·HCl
Quantity 5 g
Beilstein 13, 625
Merck Index 15, 10016
Solubility Information (10% in water) Clear colorless to light yellow
IUPAC Name hydrogen 4-(2-aminoethyl)phenol chloride
Formula Weight 173.65
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Tyramine hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.