missing translation for 'onlineSavingsMsg'
Learn More

Tyramine Hydrochloride, TRC

Product Code. 17953675
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17953675 5 g 5g
17913675 25 g 25g
17962895 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17953675 Supplier Toronto Research Chemicals Supplier No. TRCT8984935G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 60-19-5
Molecular Formula C8 H11 N O . Cl H
Molecular Weight (g/mol) 173.64
Synonym Phenol, 4-(2-aminoethyl)-, hydrochloride (1:1), Phenol, 4-(2-aminoethyl)-, hydrochloride (9CI), Phenol, p-(2-aminoethyl)-, hydrochloride (8CI), 2-(4-Hydroxyphenyl)ethylamine hydrochloride, 4-Hydroxy-β-phenethylamine hydrochloride, 4-Hydroxyphenethylamine hydrochloride, 4-Hydroxyphenylethylamine hydrochloride, Mydrial, Tyramine hydrochloride, Tyrosam, Uteramin, p-Tyramine hydrochloride
IUPAC Name 4-(2-aminoethyl)phenol;hydrochloride
SMILES Cl.NCCc1ccc(O)cc1

Specifikationer

Quantity 5 g
InChI Formula InChI=1S/C8H11NO.ClH/c9-6-5-7-1-3-8(10)4-2-7;/h1-4,10H,5-6,9H2;1H
Formula Weight 173.0607
Chemical Name or Material Tyramine hydrochloride
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.