Learn More
Fisherbrand™ Tyvek™ Cat III Coverall
Product Code. 16336525 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
16336525 2X-Large Each
16396515 Small Each
16306525 Medium Each
16316525 Large Each
16326525 X-Large Each
16346525 3X-Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 16336525
  • ONLINE EXCLUSIVE PRICE
    7.05 €
    4.28 € / Pack of 1
    Save 2.77 €
    39% Off
View all promotions
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Fisherbrand Tyvek™ Cat III Hooded Coveralls made from a soft polyethylene nonwoven fabric, offering a great balance of protection and comfort. Providing adequate Type 5 and 6 barrier protection, they remain both air and moisture vapour permeable.
  • Offers more protection than equivalent SMS garments
  • More robust than microporous films
  • Robust yet lightweight (less than 180 g/garment)
  • Elasticated face, wrists, waist and ankles
  • Antistatic treatment on the outside
  • Stitched external seams in yellow for visual identification and differentiation
TRUSTED_SUSTAINABILITY
Specifications
Product Type Hooded Coveralls
Size 2X-Large
Material Polyethylene
Color White
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 2-panel
Closure Type Zipper
Seam Type Stitched
Protection Class Category III, Type 5 and 6
Certifications/Compliance (EU) 2016/425, EN 1073-2, EN 1149-1
Size (Metric) Chest 116 to 124 cm