missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 500 Boot Covers

Product Code. p-200003643 Shop All DuPont Products
Click to view available options
Sole Type:
Anti-slip
Standard
Unit Size:
Pack of 200

Product Code. 11763083

Brand: DuPont™ D13395724

Only null left
Add to basket
Edge
Add to basket

Offers enhanced protection for body parts that are more exposed to hazardous substances. Tyvek™ 500 Boot Covers are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.

  • Knee-length overboot available in white.
  • Fixation ties.
  • Available in one size.
  • With elasticated top and stitched internal seams.
  • Permeable to both air and water vapour, yet repels water-based liquids and aerosols.
  • Excellent barrier against fine particles and fibers (down to 1 micron in size).
  • Ultra-low-linting and antistatically treated.
  • Available with standard or slip-retardant sole.
  • Applications include:
    • Pharmaceutical Handling
    • Chemical Protection
    • Lead and Asbestos Abatement/Remediation
    • General Maintenance/Operations
    • Spray Painting
    • General Clean-up
TRUSTED_SUSTAINABILITY

Specifications

Size Universal
Material HDPE
Color White
Disposable Single-use
Sterility Sterile
Closure Type Elastic with Ties
Sole Type Standard
Certifications/Compliance PPE Category I, EN 1149-5
Packaging Quantity 200 Pk.
Height (English) 18.9 in.
Height (Metric) 480 mm
Resistance Fluid
Product Type Boot Covers
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.