Learn More
DuPont™ TYVEK™ 500 HP Model TY178
Product Code. 30305955
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 25
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30305955 Small Pack of 25
30305932 Medium Pack of 25
30305897 Large Pack of 25
30305946 X-Small Pack of 25
30305970 2X-Large Pack of 25
30305941 3X-Large Pack of 25
30305921 4X-Large Pack of 25
30305956 5X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30305955 Lieferant DuPont™ Lieferanten-Nr. D15573147
  • ONLINE EXCLUSIVE PRICE
    346.65 €
    322.45 € / Pack of 25
    Save 24.20 €
    6% Off
Alle Werbeaktionen anzeigen
Estimated Shipment: 01-03-2027
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
missing translation for 'validMainframeMsg'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Harness protection coverall with four D-Ring sleeves attached to the garment. Ergonomic, protective design.
  • Workers operating with full-body harness in environments where hazardous particle or light liquid aerosol hazards are present, previously had to compromise between efficient personal protection and protecting their full-body harness against external contamination.
  • The new Tyvek™ 500 HP TY178S HP garment offers a unique solution - protection of both the wearer and the full-body harness - by providing a barrier between the user and the particle or light liquid aerosol hazards in the environment. Designed to be worn over a full-body harness, the innovative, patent-pending garment design allows workers to access and utilize multiple D-Rings on their full-body harness while providing an ideal balance of protection, durability, and comfort.
  • This new TY178S HP model has four attached sleeves for use with front, back and side D-Rings of a full-body harness. The off-centered zipper entry allows accommodating the front D-Ring sleeve for front D-Ring connection. Compatible with variety of full-body harness models, this garment may be used for a wide range of applications including painting, cleaning, maintenance, and more.
  • In addition to its innovative design, this garment also offers an ideal balance of protection and durability thanks to the unique Tyvek™ fabric. It is also extremely lightweight offering high level of comfort and mobility. The garment is low linting (suitable also for paint applications). Silicone non-added.
TRUSTED_SUSTAINABILITY
Spezifikation
Product Type Collared Coveralls
Size Small
Gender Unisex
Material HDPE
Color White
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Front Zipper with Flap
Seam Type Stitched
Sterility Non-sterile
Resistance Liquid
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.