missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 500 Shoe Covers with Antislip

Product Code. p-200003642 Shop All DuPont Products
Click to view available options
Size:
36 to 42
42 to 46
Unit Size:
Pack of 200
This item is not returnable. View return policy

Product Code. 11793113

Brand: DuPont™ D13398565

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Tyvek™ 500 Shoe Covers with Antislip are composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort. Offers enhanced protection for body parts that are more exposed to hazardous substances.

  • Slip-retardant sole.
  • Stitched internal seams.
  • With elasticated ankle.
  • White.
  • Antistatic treatment (EN 1149-5) - on both sides (excludes anti-slip sole).
  • Permeable to both air and water vapor, yet repels water-based liquids and aerosols.
  • Excellent barrier against fine particles and fibers (down to 1 micron in size).
  • Ultra-low-linting and antistatically treated.
  • Applications include:
    • Pharmaceutical Handling
    • Chemical Protection
    • Lead and Asbestos Abatement/Remediation
    • General Maintenance/Operations
    • Spray Painting
    • General Clean-up
TRUSTED_SUSTAINABILITY

Specifications

Size 36 to 42
Material Polyethylene
Color White
Sole Type Anti-slip
Seam Type Stitched
Certifications/Compliance (Europe) EN1149-5
Product Type Shoe Covers
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.