missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tyvek™ 800 J

Product Code. 15854993 Shop All DuPont Products
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
missing translation for 'unitSize':
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size missing translation for 'unitSize'
15854993 Small Pack of 25
15864993 Medium Pack of 25
15874993 Large Pack of 25
15884993 X-Large Pack of 25
15894993 2X-Large Pack of 25
15805003 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 15854993 missing translation for 'mfr' DuPont™ missing translation for 'supplierNo' D15441654
  • ONLINE EXCLUSIVE PRICE
    395.58 €
    340.72 € / Pack of 25
    Save 54.86 €
    13% Off
View all promotions
Estimated Shipment: 14-12-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

Tyvek™ 800 J coveralls benefit from the use of a specific 'impervious technology' to achieve an effective barrier against many low-concentrated water-based inorganic chemicals (even under pressure) and small-sized hazardous particulates, as well as oil repellency.

  • Tyvek™ 800J garments combine resistance to low-concentration, water-based, inorganic chemicals (even under pressure) with the strength and durability of Tyvek™ thanks to an innovative fabric technology and enhanced garment design. Developed to our highest standards, Tyvek™ 800J coveralls are soft, comfortable and lightweight without compromising their level of durability. They are now available with serged and over-taped seams that provide the performance of Type 3/4/5/6 garments, which have been tested to standards against pressurized liquids, heavy liquid aerosols and airborne solid particles. The coveralls provide an effective barrier against many water-based inorganic chemicals in low concentration and particles (down to 1.0 micron in size), as well as oil repellency. Tyvek™ 800J coveralls are suitable for applications such as industrial cleaning, chemical packaging and redistribution, waste treatment and disposal, and environmental remediation. Tyvek™ 800J Type 3/4/5/6 coveralls offer the following safety and comfort benefits:
    • Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B
    • Protection against infective agents (EN 14126) including resistance to penetration by blood and body fluids using synthetic blood (ISO 16603) and resistance to penetration by blood-borne pathogens using Phi-X174 bacteriophage (ISO 16604)
    • EN 1073-2 (protection against radioactive contamination)
    • Fabric and seams offer chemical permeation barrier to low concentration water-based inorganic chemicals
    • Serged and over-taped seams for protection and strength
    • Self-adhesive chin flap for tight seal of suit to the mask
    • Elastic face, wrists and ankles as well as glued-in waist elastic
    • Elastic thumb loops keep sleeves in place
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Protective Clothing
Size Small
Gender Unisex
Material Tyvek
Color White
Disposable Single-use
Waist Style Elastic (Glued-in)
Ankle Style Elastic
Neck Style Hooded
Hood Type Elastic
Closure Type Front Zipper with Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination)
Resistance Chemicals, Liquid, Particle, Water, Anti-static
Collar Type Hooded
Certifications/Compliance Certified according to Regulation (EU) 2016/425 Chemical protective clothing, Category III, Type 3-B, 4-B, 5-B and 6-B EN 14126 (barrier to infective agents), EN 1073-2 (protection against radioactive contamination) Antistatic treatment (EN 1149-5) - on inside
No. of Pockets None
Size (English) Chest 33-36 in.
Size (Metric) Chest 84 to 92 cm
Thickness 160 μm
For Use With (Application) Industrial cleaning, work at petrochemical installations, in sewers and maintenance operations.
Height (Metric) 162 to 170 cm
Weight (English) 59 g/m2
Quantity 25/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.