missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ 800 TJ198T ES
Click to view available options
Size:
X-Small
Small
Medium
Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Description
- DuPont™ Tyvek™ 800 ES. Hooded coverall available in sizes SM to 3X. White. Robust yet lightweight. Self-adhesive zipper flap. Self-adhesive chin flap for tight seal of suit to mask. Elasticated face, wrists and ankles as well as glued-in waist elastic. Thumb loops to prevent sleeves from riding up.
- Tyvek™ 800 ES garments are composed of PE laminated nonwoven, providing an ideal balance of protection, durability and comfort. They benefit from the use of a specific 'impervious technology' to achieve a high protection against biological hazards (EN 14126) including virus protection with an improved breathability. The fabric is antistatically treated.
- Applications for Tyvek™ 800 ES garments include delivering health care in high-risk situations, or where risk is unknown, in context of BSL3 or higher (Tuberculosis, Covid) with high exposure to body fluids: urgent medical help, road accidents, catastrophes.
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Large |
| Gender | Unisex |
| Material | HDPE |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction