Learn More
DuPont™ Tyvek 500 Jacket
Product Code. 12673690 Shop All DuPont Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12673690 Large Pack of 50
12633690 X-Large Pack of 50
12663690 2X-Large Pack of 50
12683690 Medium Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12673690 Supplier DuPont™ Supplier No. D13496057
  • ONLINE EXCLUSIVE PRICE
    426.81 €
    379.99 € / Pack of 50
    Save 46.82 €
    10% Off
View all promotions
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
Offers enhanced protection for body parts that are more exposed to hazardous substances. DuPont™ Tyvek™ 500 Jacket with Hood is composed of flash spun high density polyethylene, providing an ideal balance of protection, durability and comfort.
  • Specially designed for use with Tyvek™ apparel
  • Flash-spun HDPE construction
  • Stitched internal seams
  • Front zip closure
  • Elasticated face and wrists
  • White
  • PPE Category I
  • Antistatic treatment (EN 1149-5) - on both sides
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Material HDPE
Color White
Closure Type Zipper
Certifications/Compliance EN 1149-5
Seam Type Stitched (Internal)
Product Type Hooded Chemical-resistant Jacket
For Use With (Application) Pharmaceutical Handling, Chemical Protection, Lead and Asbestos Abatement/remediation, General Maintenance/operations, Spray Painting and General Clean-up, Amongst Many Others