Learn More
DuPont™ Tyvek™ IsoClean™ Gown
Product Code. 16498353 Shop All DuPont Products
Change view
Click to view available options
Unit Size:
Pack of 30
Size:
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size Size
16498353 Pack of 30 -
16408403 Pack of 30 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16498353 Supplier DuPont™ Supplier No. D15531630
  • ONLINE EXCLUSIVE PRICE
    268.58 €
    189.08 € / Pack of 30
    Save 79.50 €
    29% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Protective gown with bound neck.
  • Not clean-processed and not sterilized
  • Bound ties
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments Lab Coat
Color White
Cuff Style Knit
Model IC702S00
Seam Type Serged