missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyvek™ Isoclean™ Hooded Coverall IC 193 B WH DS
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
320.44 EUR valid until 2024-06-30
Use promo code "21674" to get your promotional price.
Use promo code "21674" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Clean-processed and gamma-sterilized
- The hood fits a medical mask and has attached ties
- Provides an inherent barrier to particles, microorganisms and non-hazardous water-based light liquid splash
- Breathable and low linting
- The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas
- Typically used in cleanrooms within the biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry, as well as in other critical or controlled environments
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Product Type | Cleanroom Coveralls |
Size | Small |
Gender | Unisex |
Material | HDPE |
Color | White |
Disposable | Single-use |
Waist Style | Elastic at Back |
Ankle Style | Attached Boots |
Cuff Style | Elastic |
Closure Type | Zipper with Storm Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'