Learn More
DuPont™ Tyvek™ IsoClean™ IC 270B WS Lab Coats, Sterile, Double Bagged
BIOPHARMA
Product Code. 17819323
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
3X-Large
Unit Size:
Pack of 30
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17819323 Medium Pack of 30
17899313 Small Pack of 30
17879323 Large Pack of 30
17859323 X-Large Pack of 30
17879313 - Pack of 30
17869313 3X-Large Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17819323 Supplier DuPont™ Supplier No. D15560760
  • ONLINE EXCLUSIVE PRICE
    408.07 €
    277.87 € / Pack of 30
    Save 130.20 €
    31% Off
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
Gamma-sterilized lab coats with bound neck are asepcially folded and feature bound internal seams, tunnelled at wrists, and a front snap closure.
  • Clean-processed and gamma-sterilized
  • Bound internal seams
  • Tunnelled elastication at wrists
  • Front snap closure for easy donning and doffing
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made of high density polyethylene using a patented flash spinning process Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Tyvek™ IsoClean™ is also breathable and low linting.
Tyvek™ IsoClean™ accessories (option code WS) have been sterilized by gamma- irradiation. They are folded to aid aseptic donning and packaged in an ISO class 5 cleanroom. All DuPont™ Tyvek™ IsoClean™ sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag. The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas. The accessories are individually packed and grouped together in an outer bag.
Garments and accessories made of sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments Lab Coat
Size Medium
Gender Unisex
Material Tyvek
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Sterile
Resistance Particulates, Light Liquid Splash
No. of Pockets None
Latex Free Yes