Learn More
DuPont™ Tyvek™ IsoClean™ IC 270B WS Lab Coats, Sterile, Double Bagged

Description
- Clean-processed and gamma-sterilized
- Bound internal seams
- Tunnelled elastication at wrists
- Front snap closure for easy donning and doffing
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made of high density polyethylene using a patented flash spinning process Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Tyvek™ IsoClean™ is also breathable and low linting.
Tyvek™ IsoClean™ accessories (option code WS) have been sterilized by gamma- irradiation. They are folded to aid aseptic donning and packaged in an ISO class 5 cleanroom. All DuPont™ Tyvek™ IsoClean™ sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag. The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas. The accessories are individually packed and grouped together in an outer bag.
Garments and accessories made of sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
Specifications
Specifications
| Product Type | Controlled Environments Lab Coat |
| Gender | Unisex |
| Material | Tyvek |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Sterile |
| Resistance | Particulates, Light Liquid Splash |
| No. of Pockets | None |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.