Learn More
DuPont™ Tyvek™ IsoClean™ IC 458B WS Boot Cover, Sterile, Double Bagged
BIOPHARMA
Product Code. 17809333
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17809333 Medium Pack of 100
17829333 Small Pack of 100
17839333 Large Pack of 100
17849333 X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 17809333 Supplier DuPont™ Supplier No. D15560755
  • ONLINE EXCLUSIVE PRICE
    293.56 €
    235.67 € / Pack of 100
    Save 57.89 €
    19% Off
View all promotions
Estimated Shipment: 03-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Gamma-sterilized boot covers are aseptically folded and feature covered elasticated leg opening, ankle ties, slip-retardant sole, and bound internal seams.
  • Gamma-sterilized
  • Bound internal seams
  • Tunnelled elastication at shin and ankles
  • Ankle ties
  • Gripper™ soles provide enhanced slip-retardance and robustness
Tyvek™ IsoClean™ delivers an ideal balance of protection, durability and comfort. Made of high density polyethylene using a patented flash spinning process Tyvek™ IsoClean™ provides an inherent barrier to particles, microorganisms and non-hazardous light liquid splash. Tyvek™ IsoClean™ is also breathable and low linting.
Tyvek™ IsoClean™ accessories (option code WS) have been sterilized by gamma- irradiation. They are folded to aid aseptic donning and packaged in an ISO class 5 cleanroom. All DuPont™ Tyvek™ IsoClean™ sterile accessories (option code WS) are packed in a dual barrier packaging system, consisting of an inner and outer easy tear cleanroom bag. The packaging system serves as a key element for contamination risk reduction when transferring apparel into clean areas. The accessories are individually packed and grouped together in an outer bag.
Garments and accessories made of sterile Tyvek™ IsoClean™ are typically used in cleanrooms within the hospital pharmacies, biotech, pharmaceutical, medical device manufacturing, food processing, cosmetics industry as well as in other critical or controlled environments.
TRUSTED_SUSTAINABILITY
Specifications
Size Medium
Material Tyvek
Latex Free Yes
Color White
Disposable Single-use
Sterility Sterile
Closure Type Elastic Top with Ankle Ties
Sole Type Anti-skid
Resistance Dry Particulates, Splash
Product Type Controlled Environments Boot Covers